Learn More
Thomas Scientific 50 mL SuperClear Centrifuge Tubes, 50 per Bag, Non-Sterile, 500/cs
SDP
Catalog No. NC1790035
Encompass_Preferred
SuperClear tubes offer you a standard of performance not available from other brands of centrifuge tubes. These tubes are made from a unique medical grade resin that is far more durable than commodity grade resins used in most other brands of tubes. This high quality resin makes it possible for you to spin these tubes at g-forces up to 233% greater than other tubes. So you can finally do those extractions you did in polycarbonate tubes before.
Catalog No. NC1790035 Supplier Thomas Scientific Supplier No. 2610R48
SuperClear tubes offer you a standard of performance not available from other brands of centrifuge tubes. These tubes are made from a unique medical grade resin that is far more durable than commodity grade resins used in most other brands of tubes. This high quality resin makes it possible for you to spin these tubes at g-forces up to 233% greater than other tubes. So you can finally do those extractions you did in polycarbonate tubes before.