Learn More
New England Biolabs, Inc. Supercoiled DNA Ladder – 0.1 ml
SDP
Catalog No. 504414
Encompass
The Supercoiled DNA ladder contains 9 proprietary supercoiled plasmids, ranging in size from 2 to 10 kb, that are suitable for use as supercoiled molecular weight standards for agarose electrophoresis. The 5 kb plasmid has an increased intensity to serve as a reference band.
Catalog No. 50-441-4 Supplier New England Biolabs, Inc. Supplier No. N0472S
The Supercoiled DNA ladder contains 9 proprietary supercoiled plasmids, ranging in size from 2 to 10 kb, that are suitable for use as supercoiled molecular weight standards for agarose electrophoresis. The 5 kb plasmid has an increased intensity to serve as a reference band.