Learn More
Agilent Technologies SUPERCOS I VECTOR KIT
Supplier:  Agilent Technologies 251301
Encompass
SuperCos I Vector has Cloning capacity of 30-42 kb.It is used for generating cosmid libraries and Ideal for high-resolution restriction mapping and rapid chromosomal walking.
Catalog No. 50-125-437