Learn More
Creative Biogene SUPERCULT CANINE MESENCHYM 1K
SDP
Catalog No. NC3803536
Encompass
SuperCult Canine Mesenchymal Stem Cell Growth Medium is a complete medium designed for the culture of Canine Mesenchymal Stem Cells. It was tested and optimized with Canine Mesenchymal Stem Cell culture in vitro. It is formulated for use with 5 percent CO2 and 95 percent air in a humidified incubator.
Catalog No. NC3803536 Supplier Creative Biogene Supplier No. CM00809L
SuperCult Canine Mesenchymal Stem Cell Growth Medium is a complete medium designed for the culture of Canine Mesenchymal Stem Cells. It was tested and optimized with Canine Mesenchymal Stem Cell culture in vitro. It is formulated for use with 5 percent CO2 and 95 percent air in a humidified incubator.