Learn More
Creative Biogene SUPERCULT FELINE MESENCHYMAL
SDP
Catalog No. NC3803525
Encompass
SuperCult Feline Mesenchymal Stem Cell Growth Medium is a complete medium designed for the culture of Feline Mesenchymal Stem Cells. It was tested and optimized with Feline Mesenchymal Stem Cell culture in vitro. It is formulated for use with 5 percent CO2 and 95 percent air in a humidified incubator.
Catalog No. NC3803525 Supplier Creative Biogene Supplier No. CM00810L
Add to Cart
Add to Cart
SuperCult Feline Mesenchymal Stem Cell Growth Medium is a complete medium designed for the culture of Feline Mesenchymal Stem Cells. It was tested and optimized with Feline Mesenchymal Stem Cell culture in vitro. It is formulated for use with 5 percent CO2 and 95 percent air in a humidified incubator.