Learn More
World Precision Instrument SuperCut Spring Scissors, 12.5cm, Straight
SDP
Catalog No. 502521376
Encompass
Our SuperCut Spring Scissors are surgical instruments for making small, precision cuts. They are 12.5cm long, and fit comfortably into your hand. SuperCut scissors have one edge that is micro-serrated and one that is honed razor-sharp. The micro-serrations hold the tissue firmly when the sharp blade gives a clean cut. They are often used for cutting dense or fibrous tissues. These have straight or curved tips. The spring mechanism that allows for smooth, precise cutting action, which reduces the risk of tissue damage or trauma. The spring scissors also help to mitigate hand fatigue, because they spring back naturally after you make your cut. The spring handled surgical scissors are preferred by left-handed surgeons, because they can be used ambidextrously. This gives the left-handed surgeon a more natural cutting action, which means the surgeon has greater control of the tip and the cutting movement.
Catalog No. 50-252-1376 Supplier World Precision Instrument Supplier No. 501924
Add to Cart
Add to Cart
Our SuperCut Spring Scissors are surgical instruments for making small, precision cuts. They are 12.5cm long, and fit comfortably into your hand. SuperCut scissors have one edge that is micro-serrated and one that is honed razor-sharp. The micro-serrations hold the tissue firmly when the sharp blade gives a clean cut. They are often used for cutting dense or fibrous tissues. These have straight or curved tips. The spring mechanism that allows for smooth, precise cutting action, which reduces the risk of tissue damage or trauma. The spring scissors also help to mitigate hand fatigue, because they spring back naturally after you make your cut. The spring handled surgical scissors are preferred by left-handed surgeons, because they can be used ambidextrously. This gives the left-handed surgeon a more natural cutting action, which means the surgeon has greater control of the tip and the cutting movement.