Learn More
Sigma Aldrich Fine Chemicals Biosciences SuperdexR 200 Increase HiS
Catalog No. 502908826
Encompass
If you like our new-generation Superdex Increase and Superose Increase size exclusion chromatography (SEC) columns but need to purify larger volumes of up to 8.5 mL we can pack the resin for you in HiScale columns.High-resolution SEC columns-small particle size and narrow particle size distribution of the packed resin ensures highly pure protein..nbspHiScale columns packed with SEC resins on demand please request a quote..nbspHiScale columns are packed on demand for you with Superdex 200 Increase Superdex 75 Increase and Superose 6 Increase SEC (gel filtration) resins. The columns are available in two sizes 16 x 400 mm and 26 x 400 mm. These two column sizes enable purification of sample volumes up to 3.2 and 8.5 mL respectively.
Catalog No. 50-290-8826 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE29321905
Add to Cart
Add to Cart
Request a Quote
If you like our new-generation Superdex Increase and Superose Increase size exclusion chromatography (SEC) columns but need to purify larger volumes of up to 8.5 mL we can pack the resin for you in HiScale columns.High-resolution SEC columns-small particle size and narrow particle size distribution of the packed resin ensures highly pure protein..nbspHiScale columns packed with SEC resins on demand please request a quote..nbspHiScale columns are packed on demand for you with Superdex 200 Increase Superdex 75 Increase and Superose 6 Increase SEC (gel filtration) resins. The columns are available in two sizes 16 x 400 mm and 26 x 400 mm. These two column sizes enable purification of sample volumes up to 3.2 and 8.5 mL respectively.