Learn More
Sigma Aldrich Fine Chemicals Biosciences SuperdexR SuperdexR75 Prep
Catalog No. 502904905
Encompass
Combines the high selectivity of a Sephadex matrix with the chemical and physical stability of crosslinked agarose.
Catalog No. 50-290-4905 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S665725ML
Combines the high selectivity of a Sephadex matrix with the chemical and physical stability of crosslinked agarose.