Learn More
MSC Fibre-Metal Hard Hat, Cap Style, HDPE, 8-Point Suspension, 1 ea
Catalog No. 5000508492
Encompass
This cap-style, non-vented hard hat is engineered with a high-density polyethylene (HDPE) thermoplastic shell designed to withstand high temperatures in demanding industrial environments. It features a smooth crown design and is equipped with an 8-point SuperEight™ suspension system that distributes impact energy to stabilize and balance the shell. The unit includes a wheel-ratchet headband for easy adjustment and a secure, head-hugging fit.
  • Eight load-bearing suspension points provide superior balance and stability
  • Thermoplastic shell construction offers reliable heat resistance
  • Integrated 360° padded sweatband enhances user comfort during extended wear
  • Wheel-ratchet adjustment mechanism allows for quick, precise sizing
  • Includes pre-assembled attachment blocks for rapid accessory mounting
  • Meets ANSI/ISEA Z89.1, Type I, Class E and Class C requirements
Catalog No. 50-005-08492 Supplier MSC Supplier No. 64302383
This cap-style, non-vented hard hat is engineered with a high-density polyethylene (HDPE) thermoplastic shell designed to withstand high temperatures in demanding industrial environments. It features a smooth crown design and is equipped with an 8-point SuperEight™ suspension system that distributes impact energy to stabilize and balance the shell. The unit includes a wheel-ratchet headband for easy adjustment and a secure, head-hugging fit.
  • Eight load-bearing suspension points provide superior balance and stability
  • Thermoplastic shell construction offers reliable heat resistance
  • Integrated 360° padded sweatband enhances user comfort during extended wear
  • Wheel-ratchet adjustment mechanism allows for quick, precise sizing
  • Includes pre-assembled attachment blocks for rapid accessory mounting
  • Meets ANSI/ISEA Z89.1, Type I, Class E and Class C requirements