Learn More
Trajan Scientific SUPERFLEX SYRINGE 10FX FN 10UL
SDP
Catalog No. 502530404
Encompass_Preferred
SGE Microvolume Manual Syringes, Connection Type: Fixed, Needle Gauge: 26 G, Needle Point Style: Bevel, Microvolume Manual Syringe; 10FX, Length Needle: 50 mm, 2 in., Volume: 10 uL, Diameter Outer: 0.47 mm, Length: 2 in., 50 mm, Borosilicate Glass
Catalog No. 50-253-0404 Supplier Trajan Scientific Supplier No. 002100
SGE Microvolume Manual Syringes, Connection Type: Fixed, Needle Gauge: 26 G, Needle Point Style: Bevel, Microvolume Manual Syringe; 10FX, Length Needle: 50 mm, 2 in., Volume: 10 uL, Diameter Outer: 0.47 mm, Length: 2 in., 50 mm, Borosilicate Glass