Learn More
Trajan Scientific SUPERFLEX SYRINGE FN 5FX
SDP
Catalog No. 502530419
Encompass_Preferred
Trajan Standard Plunger Protection Syringes, Length Needle: 50 mm, 1.96 in., Needle Gauge: 26 G, Volume: 5 uL, Connection Type: Fixed, Diameter Outer Needle: 0.018 in., 0.47 mm, Model: 5FX, Needle Point Number: 2, Needle Point Style: Bevel, Quantity: Each, Microvolume Syringe
Catalog No. 50-253-0419 Supplier Trajan Scientific Supplier No. 001100
Trajan Standard Plunger Protection Syringes, Length Needle: 50 mm, 1.96 in., Needle Gauge: 26 G, Volume: 5 uL, Connection Type: Fixed, Diameter Outer Needle: 0.018 in., 0.47 mm, Model: 5FX, Needle Point Number: 2, Needle Point Style: Bevel, Quantity: Each, Microvolume Syringe