Learn More
Electron Microscopy Sciences SuperFrost™ Clipped Corner
SDP
Supplier:  Electron Microscopy Sciences 7188201
Encompass_Preferred
Designed specially for the Sysmex SP-100 slide prep unit and recommended for the Diagnostics. The 45 degree clipped corner slide offers exceptional cleanliness and resistance to sticking. The tight tolerances for the length and width help to reduce accumulation of glass chips within the instrument. The pre-cleaned slide is 25 x 75 mm and approximately 1 mm thick. Ideal for technicians, who routinely prepare large number of slides.
Catalog No. 50-949-399