Learn More
Fisherbrand™ Superfrost™ Plus Microscope Slides PROMO
Catalog No. 22037246S24 Shop All Fisherbrand Products
Change view
Click to view available options
Corner Style:
Clipped
Regular
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Corner Style
22-037-246S24 Clipped
12-550-15S24 Regular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-037-246S24
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Superfrost™ Plus Microscope Slides are an ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears.
  • Maintain the same features as regular Superfrost™ slides with a special treatment process that electrostatically adheres tissue sections and cells to the glass without the need for adhesives or protein coatings
  • Treatment virtually eliminates tissue loss during staining
  • Saves hours of slide preparation
  • Chemical-resistant, white marking surface
  • 3/4 in. Superfrost™ tab
Specifications
Length (Metric) 75 mm
Width (Metric) 25 mm
Frosted 1 End
Material Glass
Type Microscope Slide
Color White
Corner Style Clipped
Quantity 72 Slides/Pk., 2 Pk./Cs.