Learn More
Epredia™ SuperFrost™ Plus Slides
Catalog No. 6776214CS Shop All Epredia Products
Change view
Click to view available options
Quantity:
1440/Cs.
Catalog No. Quantity
67-762-14CS 1440/Cs.
1 options
Catalog No. 67-762-14CS Supplier Epredia™ Supplier No. 6776214CS
Add to Cart
Add to Cart
Epredia™ Superfrost™ Plus Slides are positively charged for improved cellular adhesion.
  • Electrostatically attract tissue sections and cytology preparations, binding them to the slide
  • Eliminate the need for special adhesives or protein coatings
  • No background staining
  • Available in a variety of colors
Specifications
Frosted Yes
Type Microscope Slide
Color White
Quantity 1440/Cs.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov