Learn More
Epredia™ Superfrost™ Plus Adhesion Control Microscope Slide
Catalog No. 22042942 Shop All Epredia Products
Change view
Click to view available options
Description:
Extended Superfrost Tab
Standard Superfrost Tab
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
22-042-942 Extended Superfrost Tab
22-042-941 Standard Superfrost Tab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-042-942 Supplier Epredia™ Supplier No. I8446PLUS001
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Histology Superfrost Slide with designated area for known control sample. Available in Standard, or Extended Tab for automated and manual staining.
  • Superfrost Plus adhesion coating
  • 75 x 25 x 1 mm
  • Frosted control box outline for placing control samples
  • Available in Standard, or Extended Tab
Specifications
No. per Pack 144
Coating Superfrost Plus Adhesion
Description Extended Superfrost Tab
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov