Learn More
World Precision Instrument Cyanoacrylate Adhesive
SDP
Catalog No. 50822118
Encompass
Cyanoacrylate adhesives have been on the market since 1958. Most industrial or household grade cyanoacrylate is made of shorter alkyl chain derivatives such as methyl cyanoacrylate (WPI's #7341). They are very useful for temporarily holding tissues such as mounting specimens for microtome sectioning. This cyanoacrylate is a low viscosity adhesive.
Catalog No. 50-822-118 Supplier World Precision Instrument Supplier No. 7341
Cyanoacrylate adhesives have been on the market since 1958. Most industrial or household grade cyanoacrylate is made of shorter alkyl chain derivatives such as methyl cyanoacrylate (WPI's #7341). They are very useful for temporarily holding tissues such as mounting specimens for microtome sectioning. This cyanoacrylate is a low viscosity adhesive.