Learn More
ATHENA ES Superior Broth™ 500g
Catalog No. NC2132183
Encompass_Preferred
Superior Broth™, Superior Prime Broth™ and new Superior Primeolate™, are proprietary media formulations developed by AthenaES™ for improved expression of recombinant proteins in E. coli. The formulations are complex, supplying a nitrogen source, vitamins, and moderate glucose levels. The media are buffered at pH 7.2 ± 0.2 to prevent acidification of the medium during metabolism of the glucose. Expression of recombinant proteins should be induced at cell densities twice that of LB Broth. Cell yields are typically about 3 to 4 times that of LB Broth. Superior Broth™ shows the highest plasmid copy numbers among the AthenaES™ family of media. This medium is an excellent choice for large-scale plasmid preparations. Biomass yields will depend on the process and the strain employed.
This item is not returnable. View return policy
Catalog No. NC2132183 Supplier ATHENA ES Supplier No. 0105
Superior Broth™, Superior Prime Broth™ and new Superior Primeolate™, are proprietary media formulations developed by AthenaES™ for improved expression of recombinant proteins in E. coli. The formulations are complex, supplying a nitrogen source, vitamins, and moderate glucose levels. The media are buffered at pH 7.2 ± 0.2 to prevent acidification of the medium during metabolism of the glucose. Expression of recombinant proteins should be induced at cell densities twice that of LB Broth. Cell yields are typically about 3 to 4 times that of LB Broth. Superior Broth™ shows the highest plasmid copy numbers among the AthenaES™ family of media. This medium is an excellent choice for large-scale plasmid preparations. Biomass yields will depend on the process and the strain employed.