Learn More
Supertek™ Superior Chromatic Set of 13 Tuning Forks
GREENER_CHOICESCI_ED
Catalog No. S81832 Shop All Supertek Scientific Products
Change view
Click to view available options
Includes:
13 Tuning Forks and 1 Case
Catalog No. Includes
S81832 13 Tuning Forks and 1 Case
1 options
This item is not returnable. View return policy
Catalog No. S81832 Supplier Supertek™ Supplier No. PH36128C/2
Add to Cart
Add to Cart
This item is not returnable. View return policy
Supertek™ Superior Chromatic Set of 13 Tuning Forks are tuned to Bach's well-tempered musical scale. Made of heavy gauge aluminum. The kit comes in an attractive and practical foam lined wooden storage case.
  • Includes 1 each of:
    • 261.63 Hz 'Middle C'
    • 277.18 Hz 'C Sharp'
    • 293.7 Hz 'D'
    • 311.13 Hz 'D Sharp'
    • 329.63 Hz 'E'
    • 349.23 Hz 'F'
    • 369.99 Hz 'F#'
    • 392 Hz 'G'
    • 415.3 Hz 'G Sharp'
    • 440 Hz 'A'
    • 468.16 Hz 'A Sharp'
    • 493.88 Hz 'B'
    • 523.26 Hz 'C'
  • Each tuning fork is calibrated to within ± 1 vibration per second.
  • Kit comes in an attractive and practical foam lined wooden storage case.
Specifications
Product Type Tuning Fork
Includes 13 Tuning Forks and 1 Case
Frequency 261.63, 277.18, 293.7, 311.13, 329.63, 349.23, 369.99, 392, 415.3, 440, 468.16, 493.88, 523.26 Hz