Learn More
Superior Glove™ Aramid Fiber Stockinette Cut Resistant Sleeves
Catalog No. 17400170
Change view
Click to view available options
With thumb hole
Catalog No.
17-400-170 With thumb hole
1 options
This item is not returnable. View return policy
Catalog No. 17-400-170 Supplier Superior Glove™ Supplier No. KKWC10TH
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Construction 2-ply Aramid™ knit
Material Dupont™ Kevlar™ aramid fibers
Cuff Style Knit
For Use With (Application) Metal handling, automotive, Metal fabrication, welding
Includes Thumb hole
Length (English) 10 in.
Length (Metric) 25.4 cm