Learn More
Superior Glove™ Aramid Fiber Stockinette Cut-Resistant Sleeves
Catalog No. 17400169
Change view
Click to view available options
Length (English):
10 in.
Catalog No. Length (English)
17-400-169 10 in.
1 options
This item is not returnable. View return policy
Catalog No. 17-400-169 Supplier Superior Glove™ Supplier No. KKWC10
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Construction 2-ply Aramid™ knit
Material Dupont™ Kevlar™ aramid fibers
Cuff Style Knit
For Use With (Application) Metal handling, automotive, Metal fabrication, welding
Length (English) 10 in.
Length (Metric) 25.4 cm