Learn More
MSC Superior Glove Cut-Resistant Gloves, Nylon/Stainless-Steel, ANSI Cut A3, 1 Pair
Catalog No. 5000686663
Encompass
These gloves provide economical cut protection with a combination of nylon and fine-gauge steel. Designed for enhanced dexterity and comfort, they are ideal for handling sharp steel or glass.
  • Economical cut protection with nylon and fine-gauge steel
  • Extra-long (3 ¾ in (9 cm)) knit wrists for wrist protection and snug fit
  • Lightweight, lint-free nylon for better dexterity and touch sensitivity
  • Features 13-gauge knit for comfort and flexibility
  • Coated with foam nitrile on palms for grip
  • ANSI Cut A3 level protection
  • ANSI Abrasion Level 3
  • ANSI Puncture Level 3
Catalog No. 50-006-86663 Supplier MSC Supplier No. 66137621
These gloves provide economical cut protection with a combination of nylon and fine-gauge steel. Designed for enhanced dexterity and comfort, they are ideal for handling sharp steel or glass.
  • Economical cut protection with nylon and fine-gauge steel
  • Extra-long (3 ¾ in (9 cm)) knit wrists for wrist protection and snug fit
  • Lightweight, lint-free nylon for better dexterity and touch sensitivity
  • Features 13-gauge knit for comfort and flexibility
  • Coated with foam nitrile on palms for grip
  • ANSI Cut A3 level protection
  • ANSI Abrasion Level 3
  • ANSI Puncture Level 3