Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ CutBan™ Stockinette Cut-Resistant Sleeves

Catalog No. 17400152
Change view
Click to view available options
Length (English):
12 in.
18 in.
22 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
17-400-152 22 in.
17-400-151 18 in.
17-400-150 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-400-152 Supplier Superior Glove™ Supplier No. KPW22
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Construction 2-ply, tubular knitting with high tenacity filament Yarn
Material Yarn
Color Black
Cuff Style Knit
For Use With (Application) Metal handling, automotive, Metal fabrication
Length (English) 22 in.
Length (Metric) 55.8 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.