Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Dexterity™ Cut-Resistant String Knit Gloves, Polyurethane Coating

Catalog No. 17400190
Change view
Click to view available options
Size:
8
9
10
11
12
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-400-190 12
17-400-186 8
17-400-188 10
17-400-189 11
17-400-187 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-400-190 Supplier Superior Glove™ Supplier No. S13KFGPU12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cut-resistant gloves feature composite 13-gauge knit and polyurethane palm coating for secure grip and tactility.

  • Composite fine-gauge knit is highly dexterous
  • Cut-resistant, abrasion resistant, puncture resistant
  • Polyurethane palm coating provides secure grip and tactility
  • Ideal for glass handling, non-marring gloves will not leave residue, smudges or fingerprints
  • Dipped over an ergonomic hand form for a natural, comfortable fit

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) CE
Coating Material Polyurethane
Color Cream, Gray
Cut Resistance Rating ANSI A4
Hand Specific
Material Aramid Fiber, Silica
Product Type Cut-resistant Gloves
Size 12
EN388 Standard EN 388:4X31B
For Use With (Application) automotive, light Metal stamping, fabrication, small parts handling assembly, Glass handling, sheet Metal work
Glove Cuff Style Knit
Model S13KFGPU/12
Puncture Rating ANSI/ISEA 105-16 Level 2
Series Dexterity™
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.