Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Dexterity™ Cut-Resistant String Knit Gloves, Nitrile Coating

Catalog No. 17401200
Change view
Click to view available options
Size:
6
7
8
9
11
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-200 11
17-400-191 6
17-400-192 7
17-400-193 8
17-400-194 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-401-200 Supplier Superior Glove™ Supplier No. S13KFNT11
This item is not returnable. View return policy

Aramid fiber composite string knit gloves provides ANSI Level A3 cut-resistance. Nitrile coating makes this glove ideal for a variety of dry grip applications.

  • Combination of Kevlar™ and composite filament fiber provides 360° cut protection
  • Nitrile palm coating offers secure dry grip and durability
  • Ergonomically designed for unsurpassed comfort and dexterity
  • Cut, abrasion and puncture resistant

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile Foam
Color Cream, Black
Cut Resistance Rating ANSI A3
Material Dexterity
Product Type Cut-resistant Gloves
Size 11
For Use With (Application) Automotive, light metal stamping, small parts handling, maintenance jobs, HVAC, glass
Glove Cuff Style Knit
Model S13KFNT/11
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Dexterity™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.