Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Dexterity™ Winter-Lined Nylon Gloves with PVC Palms

Catalog No. 17401295
Change view
Click to view available options
Size:
X-Large
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-295 2X-Large
17-401-294 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-401-295 Supplier Superior Glove™ Supplier No. SNTAPVC/XX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Micropore PVC coating enhances grip capabilities and remains flexible in sub-zero temperatures down to -5°C. Gloves are treated with a water repellent formula for quick-drying, all day comfort.

  • Micropore PVC coating remains flexible in sub-zero temperatures down to -5°C
  • Coating enhances grip capabilites in wet and dry conditions
  • Fleece lining is soft and warm for protection during cold applications
  • Quick-drying and lightweight for all-day comfort
  • Treated with water repellent formula

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material PVC
Color Red
Cut Resistance Rating ANSI A3
Hand Specific
Material Nylon
Size 2X-Large
For Use With (Application) Agriculture, Construction, Cold Storage, Commercial Fishing, and Utilities
Glove Cuff Style Knit
Liner Material Fleece
Series SNTAPVC
Temperature (English) Min 23°F
Temperature (Metric) Min. -5°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.