Learn More
MSC Superior Glove Glove Liners, ANSI A4 Cut, Large, 1 Pair
Catalog No. 5000770008
Encompass
These heat-resistant glove liners provide excellent protection against cut and heat hazards. Made with ParaActiv™ engineered yarn, they offer 360° ANSI A4 cut resistance and ANSI 2 heat protection, while maintaining dexterity and flexibility for a comfortable, ergonomic fit.
  • Engineered yarn provides 360° cut protection with ANSI A4 cut resistance and ANSI 2 heat protection.
  • Composite fine-gauge knit offers high dexterity and flexibility.
  • Ergonomically shaped for improved hand movement and a snug fit.
  • ANSI Level 3 abrasion resistance.
  • Composite filament fiber wrapped with para-aramid fiber for enhanced cut resistance without adding bulk.
  • Seamless knit construction.
  • Suitable for use on their own or as liners under other gloves.
Catalog No. 50-007-70008 Supplier MSC Supplier No. 13723903
These heat-resistant glove liners provide excellent protection against cut and heat hazards. Made with ParaActiv™ engineered yarn, they offer 360° ANSI A4 cut resistance and ANSI 2 heat protection, while maintaining dexterity and flexibility for a comfortable, ergonomic fit.
  • Engineered yarn provides 360° cut protection with ANSI A4 cut resistance and ANSI 2 heat protection.
  • Composite fine-gauge knit offers high dexterity and flexibility.
  • Ergonomically shaped for improved hand movement and a snug fit.
  • ANSI Level 3 abrasion resistance.
  • Composite filament fiber wrapped with para-aramid fiber for enhanced cut resistance without adding bulk.
  • Seamless knit construction.
  • Suitable for use on their own or as liners under other gloves.