Learn More
MSC Superior Glove Gloves, Waterproof, A6 Cut, Size 11, 1 Pair
Catalog No. 5000685527
Encompass
The Dexterity® S18WTFN gloves provide waterproof and windproof protection with laminated anti-slip layers for enhanced grip and cut resistance. Constructed with TenActiv™ engineered yarn, they offer 360° cut protection (ANSI A6) and a foam nitrile palm coating for a steady wet grip.
  • Waterproof and windproof protection; breathable.
  • Laminated anti-slip layers for enhanced grip and dexterity.
  • TenActiv™ engineered yarn provides 360° cut protection (ANSI A6).
  • Foam nitrile palm coating offers steady wet grip.
  • Reinforced thumb crotch for durability against wear and tear.
  • Easy care: washable with bleach or detergent.
  • Available in sizes 6-12.
  • ANSI Level 6 abrasion, A6 360° cut, and Level 4 puncture protection.
Catalog No. 50-006-85527 Supplier MSC Supplier No. 02632446
The Dexterity® S18WTFN gloves provide waterproof and windproof protection with laminated anti-slip layers for enhanced grip and cut resistance. Constructed with TenActiv™ engineered yarn, they offer 360° cut protection (ANSI A6) and a foam nitrile palm coating for a steady wet grip.
  • Waterproof and windproof protection; breathable.
  • Laminated anti-slip layers for enhanced grip and dexterity.
  • TenActiv™ engineered yarn provides 360° cut protection (ANSI A6).
  • Foam nitrile palm coating offers steady wet grip.
  • Reinforced thumb crotch for durability against wear and tear.
  • Easy care: washable with bleach or detergent.
  • Available in sizes 6-12.
  • ANSI Level 6 abrasion, A6 360° cut, and Level 4 puncture protection.