Learn More
MSC Superior Glove Goatskin Gloves, Arc Flash, A4 Cut, 1 Pair
Catalog No. 5000796609
Encompass
These goatskin gloves are designed for non-energized work in arc flash environments where liquid resistance is crucial. Constructed with flame-resistant material, they feature a ParaActiv™ engineered yarn liner for 360° cut protection. Unpadded palms allow for a steady grip and an improved sense of feel during tasks.
  • Arc flash rated level 4 with an Arc Thermal Performance Value (ATPV) of 41 cal/cm²
  • Passes ASTM D6413 vertical flame resistance test
  • ParaActiv™ engineered yarn liner provides 360° ANSI A4 cut protection
  • Provides ANSI 3 heat resistance (200°C or 392°F)
  • Treated to resist water and liquids
  • Unpadded palms for enhanced sense of feel and better grip
Catalog No. 50-007-96609 Supplier MSC Supplier No. 16554230
These goatskin gloves are designed for non-energized work in arc flash environments where liquid resistance is crucial. Constructed with flame-resistant material, they feature a ParaActiv™ engineered yarn liner for 360° cut protection. Unpadded palms allow for a steady grip and an improved sense of feel during tasks.
  • Arc flash rated level 4 with an Arc Thermal Performance Value (ATPV) of 41 cal/cm²
  • Passes ASTM D6413 vertical flame resistance test
  • ParaActiv™ engineered yarn liner provides 360° ANSI A4 cut protection
  • Provides ANSI 3 heat resistance (200°C or 392°F)
  • Treated to resist water and liquids
  • Unpadded palms for enhanced sense of feel and better grip