Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Protex™ Tapered Cut-Resistant Sleeves

Catalog No. 17400149
Change view
Click to view available options
Color:
Green
Red
White
Yellow
Length (English):
18 in.
22 in.
Size:
Small
Medium
Large
X-Large
Includes:
Elastic biceps
Thumb hole, elastic biceps
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (English) Size Includes
17-400-149 Green 22 in. X-Large Thumb hole, elastic biceps
17-400-134 Red 18 in. Small Elastic biceps
17-400-135 Yellow 18 in. Medium Elastic biceps
17-400-136 White 18 in. Large Elastic biceps
17-400-137 Green 18 in. X-Large Elastic biceps
17-400-138 Red 18 in. Small Thumb hole, elastic biceps
17-400-139 Yellow 18 in. Medium Thumb hole, elastic biceps
17-400-140 White 18 in. Large Thumb hole, elastic biceps
17-400-141 Green 18 in. X-Large Thumb hole, elastic biceps
17-400-143 Yellow 22 in. Medium Elastic biceps
17-400-144 White 22 in. Large Elastic biceps
17-400-142 Red 22 in. Small Elastic biceps
17-400-145 Green 22 in. X-Large Elastic biceps
17-400-147 Yellow 22 in. Medium Thumb hole, elastic biceps
17-400-148 White 22 in. Large Thumb hole, elastic biceps
17-400-146 Red 22 in. Small Thumb hole, elastic biceps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 17-400-149 Supplier Superior Glove™ Supplier No. KPXT22TH/X
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Construction 2-ply, flame resistant Protex™ Yarn
Size X-Large
Material Modacrylic Fiber
Color Green
Cuff Style Knit
For Use With (Application) Metal handling, automotive, Metal fabrication
Includes Thumb hole, elastic biceps
Length (English) 22 in.
Length (Metric) 55.8 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.