Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Superior Touch™ Dyneema™ Gloves, Foam Nitrile Palm

Catalog No. 17400179
Change view
Click to view available options
Size:
6
12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-400-179 6
17-400-185 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-400-179 Supplier Superior Glove™ Supplier No. S13SXGBFN6
Add to Cart
Add to Cart
This item is not returnable. View return policy

Speckled gray Dyneema™ cut-resistant gloves feature a foam nitrile palm for secure grip when working with liquids and oils.

  • Speckled gray, 13-gauge gloves
  • Made of knit with lint-free, continuous-filament Dyneema™
  • Foam nitrile improves grip by absorbing and displacing liquids and oils on smooth objects
  • Gloves are washable and can be bleached at cold temperatures only

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Nitrile Foam
Color Gray, Black
Cut Resistance Rating ANSI A2
Material Dyneema
Product Type Cut-resistant Gloves
Size 6
For Use With (Application) sharp Steel, Glass, circuit boards, automotive
Glove Cuff Style Knit
Model S13SXGBFN/6
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Superior Touch™
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.