Learn More
MSC Superior Glove Works Anti-Vibration Glove, Half-Finger, Goatskin, Small, 1 Pair
Catalog No. 5000792916
Encompass
These half-finger gloves offer anti-vibration and impact protection, designed for demanding applications such as construction, mining, and automotive work. They are constructed from durable goatskin leather and feature a wrist strap cuff for a secure fit.
  • Offers a snug fit and comfort
  • Adjustable straps support the wrist
  • Leather padded palm
  • Meets ANSI Abrasion 4 standard
Catalog No. 50-007-92916 Supplier MSC Supplier No. 90494881
These half-finger gloves offer anti-vibration and impact protection, designed for demanding applications such as construction, mining, and automotive work. They are constructed from durable goatskin leather and feature a wrist strap cuff for a secure fit.
  • Offers a snug fit and comfort
  • Adjustable straps support the wrist
  • Leather padded palm
  • Meets ANSI Abrasion 4 standard