Learn More
MSC Superior Glove Works Arc Flash Glove, X-Large, 1 ea
Catalog No. 5000768579
Encompass
These arc flash gloves are designed to provide hand protection in environments with arc flash hazards. They feature high levels of abrasion and cut resistance, ensuring durability and safety for the wearer.
  • X-Large size for comfortable fit
  • Numeric size 10
  • ANSI/ISEA abrasion resistance level 3
  • ANSI/ISEA cut resistance level A4
  • Provided as a pair
Catalog No. 50-007-68579 Supplier MSC Supplier No. 66271057
These arc flash gloves are designed to provide hand protection in environments with arc flash hazards. They feature high levels of abrasion and cut resistance, ensuring durability and safety for the wearer.
  • X-Large size for comfortable fit
  • Numeric size 10
  • ANSI/ISEA abrasion resistance level 3
  • ANSI/ISEA cut resistance level A4
  • Provided as a pair