Learn More
MSC Superior Glove Works Cold Gloves, Nylon/Cotton, Nitrile-Coated, Large, 1 Pair
Catalog No. 5000794161
Encompass
These cold gloves are designed for general purpose applications, including construction. They feature a knit cuff and a textured grip surface for enhanced handling and dexterity in various work environments.
  • General purpose glove type
  • Applications: Construction, general purpose
  • Knit cuff style
  • Full coating coverage except cuff
  • Textured grip surface for improved handling
  • Large size, numeric size 9
  • Cuff length: 2 inch
  • Wing thumb type
  • Yellow and black color
  • ANSI/ISEA abrasion resistance level 4
  • Protection against oil
  • Seamless design for comfort
Catalog No. 50-007-94161 Supplier MSC Supplier No. 56872195
These cold gloves are designed for general purpose applications, including construction. They feature a knit cuff and a textured grip surface for enhanced handling and dexterity in various work environments.
  • General purpose glove type
  • Applications: Construction, general purpose
  • Knit cuff style
  • Full coating coverage except cuff
  • Textured grip surface for improved handling
  • Large size, numeric size 9
  • Cuff length: 2 inch
  • Wing thumb type
  • Yellow and black color
  • ANSI/ISEA abrasion resistance level 4
  • Protection against oil
  • Seamless design for comfort