Learn More
MSC Superior Glove Works Cut A2 Gloves, X-Small, 12 Pair
Catalog No. 5000803217
Encompass
These gloves are engineered for robust protection against cuts, punctures, and abrasions. They feature a TenActiv, polyurethane-coated palm and fingers with a smooth grip, designed to provide excellent dexterity and tactile sensitivity for general-purpose handling in demanding environments.
  • ANSI Level 4 abrasion resistance
  • Provides 360° ANSI level A2 cut protection
  • ANSI Level 3 puncture resistance
  • Lint-free, continuous filament yarn
  • Snug fitting, breathable, and flexible
  • Ergonomically shaped
  • Polyurethane coating offers excellent dexterity and tactile sensitivity
Catalog No. 50-008-03217 Supplier MSC Supplier No. 53920161
These gloves are engineered for robust protection against cuts, punctures, and abrasions. They feature a TenActiv, polyurethane-coated palm and fingers with a smooth grip, designed to provide excellent dexterity and tactile sensitivity for general-purpose handling in demanding environments.
  • ANSI Level 4 abrasion resistance
  • Provides 360° ANSI level A2 cut protection
  • ANSI Level 3 puncture resistance
  • Lint-free, continuous filament yarn
  • Snug fitting, breathable, and flexible
  • Ergonomically shaped
  • Polyurethane coating offers excellent dexterity and tactile sensitivity