Learn More
MSC Superior Glove Works Dyneema Cold Gloves, Polyurethane, 2X-Large, 1 Pair
Catalog No. 5000676602
Encompass
These cold gloves are designed for general purpose applications, offering cut protection and textured grip. Made with Dyneema material, they are suitable for various tasks requiring dexterity and safety.
  • General purpose glove type
  • Dyneema material for glove, lining, back, and cuff
  • Polyurethane coating on palm and fingertips for enhanced grip
  • Knit cuff style with a 2 in. length
  • Fingered design with 3/4 coverage for improved dexterity
  • Seamless construction for comfort
  • Meets ANSI Puncture 3 and ANSI/ISEA Cut Level A2 standards
  • Unisex design, 2X-Large size
Catalog No. 50-006-76602 Supplier MSC Supplier No. 11960929
These cold gloves are designed for general purpose applications, offering cut protection and textured grip. Made with Dyneema material, they are suitable for various tasks requiring dexterity and safety.
  • General purpose glove type
  • Dyneema material for glove, lining, back, and cuff
  • Polyurethane coating on palm and fingertips for enhanced grip
  • Knit cuff style with a 2 in. length
  • Fingered design with 3/4 coverage for improved dexterity
  • Seamless construction for comfort
  • Meets ANSI Puncture 3 and ANSI/ISEA Cut Level A2 standards
  • Unisex design, 2X-Large size