Learn More
MSC Superior Glove Works Glove, TenActiv, 21-Gauge, ANSI A9, 1 Pair
Catalog No. 5000766337
Encompass
These gloves offer industry-leading A9 cut protection in a thin 21-gauge knit for a barehand feel. Featuring a leather-reinforced thumb crotch with TenActiv™ thread for enhanced durability and abrasion resistance, they are ideal for extreme cut-hazard tasks. The ultra-thin foam nitrile palm coating is touchscreen compatible and provides a strong grip in lightly oiled conditions.
  • Offers industry's highest ANSI A9 cut resistance with 21-gauge knit
  • Leather thumb crotch for protection against wear and tear; reinforced with TenActiv™ thread for added seam integrity
  • Ultra-thin foam nitrile palm coating provides a better grip for handling lightly oiled parts
  • Barehand feel balances both durability and flexibility
  • Touchscreen compatible to seamlessly move between tasks
  • Latex free
  • TAA compliant
Catalog No. 50-007-66337 Supplier MSC Supplier No. 89740658
These gloves offer industry-leading A9 cut protection in a thin 21-gauge knit for a barehand feel. Featuring a leather-reinforced thumb crotch with TenActiv™ thread for enhanced durability and abrasion resistance, they are ideal for extreme cut-hazard tasks. The ultra-thin foam nitrile palm coating is touchscreen compatible and provides a strong grip in lightly oiled conditions.
  • Offers industry's highest ANSI A9 cut resistance with 21-gauge knit
  • Leather thumb crotch for protection against wear and tear; reinforced with TenActiv™ thread for added seam integrity
  • Ultra-thin foam nitrile palm coating provides a better grip for handling lightly oiled parts
  • Barehand feel balances both durability and flexibility
  • Touchscreen compatible to seamlessly move between tasks
  • Latex free
  • TAA compliant