These ParaActiv™ S13KFGPU gloves offer 360° cut protection with a moderate ANSI A4 cut resistance, designed with a fine knit for a bare-hand feel. The non-marring polyurethane palm dip coating provides enhanced dry grip without leaving residue, and their ergonomic shape ensures a close, comfortable fit, making them ideal for delicate tasks such as glass handling.
- ParaActiv™ engineered yarn liner provides 360° cut protection with ANSI A4 cut resistance.
- Non-marring polyurethane palm dip coating enhances dry grip and prevents residue transfer.
- Ergonomically shaped for a natural, close, and comfortable fit.
- Ideal for glass handling as they will not leave smudges or fingerprints.
- Features 13-gauge knit construction for enhanced dexterity.
- Offers ANSI Level 4 abrasion resistance and ANSI Level 2 puncture resistance.