Learn More
MSC Superior Glove Works Gloves, ANSI A5 Cut, X-Large, 12 Pairs
Catalog No. 5000792506
Encompass
These industrial gloves are engineered with high-strength Tenactiv yarn for ANSI A5 cut protection. Featuring a high-visibility knit shell and a polyurethane coated palm, they provide excellent dexterity, tactile sensitivity, and a strong grip suitable for demanding work environments.
  • Engineered with high-strength Tenactiv yarn
  • Provides ANSI A5 cut protection
  • Features high-visibility knit shell for increased worker visibility
  • Polyurethane coating on palm and fingers offers excellent dexterity, tactile sensitivity, and strong grip
  • Lint-free shell for contaminant-free work
  • Offers ANSI 4 puncture and ANSI 4 abrasion resistance
  • Static dissipative
Catalog No. 50-007-92506 Supplier MSC Supplier No. 54805114
These industrial gloves are engineered with high-strength Tenactiv yarn for ANSI A5 cut protection. Featuring a high-visibility knit shell and a polyurethane coated palm, they provide excellent dexterity, tactile sensitivity, and a strong grip suitable for demanding work environments.
  • Engineered with high-strength Tenactiv yarn
  • Provides ANSI A5 cut protection
  • Features high-visibility knit shell for increased worker visibility
  • Polyurethane coating on palm and fingers offers excellent dexterity, tactile sensitivity, and strong grip
  • Lint-free shell for contaminant-free work
  • Offers ANSI 4 puncture and ANSI 4 abrasion resistance
  • Static dissipative