Learn More
MSC Superior Glove Works Gloves, Cut A2, Size Small, 12/Pk.
Catalog No. 5000796873
Encompass
These gloves are designed for cut, puncture, and abrasion resistance, featuring a polyester shell with a micropore nitrile coating on the palm and fingertips. They offer a textured grip for enhanced handling.
  • Micropore nitrile coating on palm and fingertips
  • Textured grip surface
  • Knit cuff style
  • ANSI/ISEA Cut Resistance Level A2
  • ANSI/ISEA Abrasion Resistance Level 3
  • ANSI/ISEA Puncture Resistance Level 2
  • Polyester primary material
  • Dyneema back, lining, and cuff material
Catalog No. 50-007-96873 Supplier MSC Supplier No. 11961059
These gloves are designed for cut, puncture, and abrasion resistance, featuring a polyester shell with a micropore nitrile coating on the palm and fingertips. They offer a textured grip for enhanced handling.
  • Micropore nitrile coating on palm and fingertips
  • Textured grip surface
  • Knit cuff style
  • ANSI/ISEA Cut Resistance Level A2
  • ANSI/ISEA Abrasion Resistance Level 3
  • ANSI/ISEA Puncture Resistance Level 2
  • Polyester primary material
  • Dyneema back, lining, and cuff material