Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Superior Touch™ Cleanroom Processed Cut-Resistant Gloves

Catalog No. 17489649
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-489-649 9
17-489-646 6
17-489-647 7
17-489-648 8
17-489-650 10
17-489-651 11
17-489-652 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-489-649 Supplier Superior Glove™ Supplier No. CS13SXPUQ9
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dyneema™ fiber gloves with polyurethane coated palms are clean processed and ideal for sharp steel, glass, food and circuit board applications.

  • Dyneema™ is lint-free and provides cut and abrasion resistance
  • Polyurethane coated palm provides secure grip in wet and oily conditions
  • Form fitting, breathable and flexible
  • Ergonomically shaped

Specifications

Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A2
Material Dyneema
Product Type Cut-resistant Gloves
Size 9
For Use With (Application) Sharp steel, glass, circuit boards, food applications
Glove Cuff Style Knit
Model CS13SXPUQ-9
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Superior Touch™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.