Learn More
AdipoGen SuperKillerTRAIL, Soluble (human) (rec.),Protein.
SDP
Supplier:  AdipoGen AG40T0002C020
Encompass_Preferred
SuperKillerTRAIL, Soluble (human) (rec.), AdipoGen Life Sciences. Protein. The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to a His-tag and a linker peptide. The active multimeric conformation is stabilized by an inserted mutation allowing an additional CC-bridge. Source: E. coli. Endotoxin content: <0.01EU/?g purified protein (LAL test; Lonza). Liquid. In 20mM HEPES, pH 7.4, 150mM NaCl, 0.005% Tween 20, 1% sucrose, 1mM DTT. Binds to human TRAIL receptors. Purity: >98% (SDS-PAGE). TNF-related apoptosis-inducing ligand (TRAIL; Apo2L;CD253; TNFSF10) is a type II transmembrane protein of about 34kDa. Like most members of the tumor necrosis factor (TNF) superfamily of cytokines TRAIL can be cleaved at the cell surface by metalloproteases to form a soluble molecule.
Catalog No. 50-464-153