Learn More
V&P Scientific Inc MAGNETIC PLATE, 96 Well, PCR, V-Bottom, 12 Bar Halbach Array
Catalog No. NC2040907
Encompass
MAGNETIC BEAD SEPARATION PLATE, 96 Well, PCR Plate, V-Bottom, 12 Bar Halbach Array, 52MGO, NdFeB Magnets, Gold Anodized Aluminum Magnet Frame, 1 Sided Pellet Location, Halbach Array, Gripper Accessible, for Half-Skirt, Not for Full-Skirt, Beads Collected to a Single Side of a Row of Wells, SLAS Footprint, No Registration Base
Catalog No. NC2040907 Supplier V&P Scientific Inc Supplier No. VP 771V1
MAGNETIC BEAD SEPARATION PLATE, 96 Well, PCR Plate, V-Bottom, 12 Bar Halbach Array, 52MGO, NdFeB Magnets, Gold Anodized Aluminum Magnet Frame, 1 Sided Pellet Location, Halbach Array, Gripper Accessible, for Half-Skirt, Not for Full-Skirt, Beads Collected to a Single Side of a Row of Wells, SLAS Footprint, No Registration Base