Learn More
Sigma Aldrich Supermag Multitube Magnetic Separator
Catalog No. 503125995 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-312-5995 1 Ea.
1 options
Catalog No. 50-312-5995 Supplier Sigma Aldrich Supplier No. OCNMM31.51EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
The SuperMag Multitube Separator is designed for easy and rapid separation of magnetic nanocrystals in small volume in as many as ten different samples simultaneously. It uses miniaturized magnets with very high magnetic fields. It weighs only about 1.23 kg with the following dimensions: 4.5 cm x 18 cm x 7.3 cm. It is completely portable and can accommodate ten pieces of 1.5-mL centrifuge tubes. It is recommended for the separation of 20 nm magnetic nanocrystals and above. Times required for separation will vary depending on the particle size and the magnetic nanocrystal concentration in the solution. The SuperMag Multitube Separator has a strong magnetic field. This product should not be used near magnets, metal, credit cards, or memory storage devices. Failure to follow this warning could lead to serious injury and loss of valuable data.Product of Ocean Nanotech, LLC
Specifications
Quantity 1 Ea.