Learn More
Sino Biological SuperNuclease (50% Glycerol) 50 kU
Catalog No. 5000696264
Encompass_Preferred
SuperNuclease is a recombined Serratia marcescens’ extracellular endonuclease. The sequence is same with Benzonase (Benzonase is a trademark of Merck KGaA, Darmstadt, Germany).
Catalog No. 50-006-96264 Supplier Sino Biological Supplier No. GMPSSNP01G50
SuperNuclease is a recombined Serratia marcescens’ extracellular endonuclease. The sequence is same with Benzonase (Benzonase is a trademark of Merck KGaA, Darmstadt, Germany).