Learn More
Sino Biological SuperNuclease
Supplier:  Sino Biological SSNP0150
Encompass_Preferred
SuperNuclease is a recombined Serratia marcescens’ extracellular endonuclease. The sequence is same with Benzonase (Benzonase is a trademark of Merck KGaA, Darmstadt, Germany).
Catalog No. NC1647647