Learn More
Sigma Aldrich Fine Chemicals Biosciences SuperoseR 6 Prep Grade HiL
Catalog No. 502908874
Encompass
HiLoad 16/600 Superose 6 prep grade (pg) is a prepacked size exclusion chromatography (SEC) column for high-resolution purification of large biomolecules and protein complexes. The column is an excellent choice for purification of sample volumes up to 5 mL..nbspConvenience and reproducibility can be used with standalone pump or a chromatography system..nbspHigh-resolution separations purification is possible across an exceptionally broad molecular-weight range..nbspScalable separations can be scaled up using Superose 6 prep grade resin..nbspÄKTA system compatible fittings (1/16) for direct connection to ÄKTA systems and other chromatography systems.Selectivity for a wide range of proteinsThis column is packed with Superose 6 pg resin which provides good resolution separations of proteins and other biomolecules according to size. The wide fractionation range of the resin enables reproducible and robust separations in the molecular weight range from 5000 up to 5 000 000.
Catalog No. 50-290-8874 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE29323952
HiLoad 16/600 Superose 6 prep grade (pg) is a prepacked size exclusion chromatography (SEC) column for high-resolution purification of large biomolecules and protein complexes. The column is an excellent choice for purification of sample volumes up to 5 mL..nbspConvenience and reproducibility can be used with standalone pump or a chromatography system..nbspHigh-resolution separations purification is possible across an exceptionally broad molecular-weight range..nbspScalable separations can be scaled up using Superose 6 prep grade resin..nbspÄKTA system compatible fittings (1/16) for direct connection to ÄKTA systems and other chromatography systems.Selectivity for a wide range of proteinsThis column is packed with Superose 6 pg resin which provides good resolution separations of proteins and other biomolecules according to size. The wide fractionation range of the resin enables reproducible and robust separations in the molecular weight range from 5000 up to 5 000 000.