Learn More
Cusabio Technology LLC superoxide dismutase 1
Catalog No. 5000051048
Encompass_Preferred
This kit detects SOD1 in serum plasma cell culture supernates and tissue homogenates offering a quantitative measurement range of 15 6 pg/mL to 1000 pg/mL with a sensitivity of 3 9 pg/mL Designed for efficient analysis it employs a sandwich ELISA methodology requiring a sample volume of 50-100 L and delivering results within 1-5 hours The assay utilizes absorbance measurement at 450 nm to ensure accurate detection and quantification of SOD1 across diverse biological samples making it suitable for research applications demanding high precision and reliability in measuring soluble superoxide dismutase 1 levels
Catalog No. 50-000-51048 Supplier Cusabio Technology LLC Supplier No. CSBEL022397RA
This kit detects SOD1 in serum plasma cell culture supernates and tissue homogenates offering a quantitative measurement range of 15 6 pg/mL to 1000 pg/mL with a sensitivity of 3 9 pg/mL Designed for efficient analysis it employs a sandwich ELISA methodology requiring a sample volume of 50-100 L and delivering results within 1-5 hours The assay utilizes absorbance measurement at 450 nm to ensure accurate detection and quantification of SOD1 across diverse biological samples making it suitable for research applications demanding high precision and reliability in measuring soluble superoxide dismutase 1 levels