Learn More
Worthington Biochemical Corporation SUPEROXIDE DISMUTASE 2MG
SDP
Catalog No. 50592359
Encompass
Chromatographically purified essentially as described by McCord and Fridovich, J. Biol. Chem., 244, 6049 (1969). A dialyzed, lyophilized powder. 9054-89-1
Catalog No. 50-592-359 Supplier Worthington Biochemical Corporation Supplier No. LS003540
Chromatographically purified essentially as described by McCord and Fridovich, J. Biol. Chem., 244, 6049 (1969). A dialyzed, lyophilized powder. 9054-89-1