Learn More
Enzo Life Sciences SOD activity kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900157
Encompass_Preferred
ELS offers a large collection of superoxide dismutase products for oxidative stress research. This is a versatile assay for the determination of SOD activity. Differentiate between the activity of three SOD isoenzymes: MnSOD, FeSOD, and Cu/ZnSOD. Obtain higher throughput sample analysis with a convenient colorimetric 96 well plate format and fast results in just 10 minutes. Alternative Name: Superoxide dismutase. Applications: Colorimetric detection, Activity assay. Contents: Microtiter plate, Standard, SOD buffer concentrate, Xanthine oxidase, Xanthine solution, Triton x-100, WST-1 reagent
Catalog No. 50-658-183