Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Superoxide Dismutase, MP Biomedicals™

Catalog No. MP219011732 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
300 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219011732 300 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219011732 Supplier MP Biomedicals™ Supplier No. 0219011732
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Catalyzes the dismutation of superoxide into oxygen and hydrogen peroxide. Superoxide Dismutase, MP Biomedicals is an antioxidant enzyme and an important defense in cells exposed to oxygen. It is a low molecular weight metalloprotein present in aerobic ce

Superoxide Dismutase (SOD) provides protection against damaging reactions with the superoxide radical anion (O2-) by catalyzing its disproportionation into oxygen and hydrogen peroxide. It competes with nitric oxide (NO) for superoxide anion (which reacts with NO to form peroxynitrite), thereby promoting the activity of NO. SOD has also been shown to suppress apoptosis in cultured rat ovarian follicles, neural cell lines, and transgenic mice.

Superoxide Dismutase plays a critical role in the defense of cells against the toxic effects of oxygen radicals. SOD from bovine erythrocytes has been used in a study to assess a kinetic model of radiation-induced inactivation of superoxide dismutase in nitrous oxide-saturated solutions. Superoxide dismutase from bovine erythrocytes has also been used in a study to investigate the possible participation of superoxide anion in the intestinal tryptophan 2,3-dioxygenase reaction.

TRUSTED_SUSTAINABILITY

Specifications

Color Pale Blue
Description Superoxide dismutase
CAS 9054-89-1
Synonym SOD; Superoxide: superoxide oxidoreductase
Content And Storage Store at -20°C; desiccate
Donor Source Bovine Erythrocytes
Form Lyophilized
For Use With (Application) Catalyst in dismutation of superoxide
Quantity 300 kU
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.